Imprimer

Anti-CTLA4 Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB26605-0.2EA 0 EUR
BIRBORB26605-0.2
Anti-CTLA4 Mouse monoclonal antibody
Anticorps
Flow Cytometry: 2 ?g

Type: Primary
Antigen: CTLA4 (cytotoxic T-lymphocyte-associated protein 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR