Anti-8-Hydroxyguanosine Goat Polyclonal Antibody
Supplier: Biorbyt
BIRBORB26686-0.2EA 0 EUR
BIRBORB26686-0.2
Anti-8-Hydroxyguanosine Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 8-Hydroxyguanosine (8-hydroxyguanosine)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now