Anti-UCHL1 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26696-0.2EA 0 EUR
BIRBORB26696-0.2
Anti-UCHL1 Mouse Monoclonal Antibody
Antibodies
Immunochemistry: 1:50-1:400; ELISA: 1:160000

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR