Anti-UCHL1 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26697-0.2EA 0 EUR
BIRBORB26697-0.2
Anti-UCHL1 Mouse Monoclonal Antibody
Antibodies
ELISA: 1:500-1:2000; Immunochemistry: 1:100-1:400

Type: Primary
Antigen: UCHL1 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR