Anti-Nicotinic alpha 4 Rec. Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26709-100EA 0 EUR
BIRBORB26709-100
Anti-Nicotinic alpha 4 Rec. Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:200

Type: Primary
Antigen: Nicotinic alpha 4 Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR