Anti-D4 Dopamine Recept Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26725-100EA 0 EUR
BIRBORB26725-100
Anti-D4 Dopamine Recept Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: D4 Dopamine Recept
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR