Anti-Nicotinic beta 4 Rec. Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26728-100EA 0 EUR
BIRBORB26728-100
Anti-Nicotinic beta 4 Rec. Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 1:100; Western Blot: 1:200

Type: Primary
Antigen: Nicotinic beta 4 Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR