Anti-NOS3 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB26756-100EA 0 EUR
BIRBORB26756-100
Anti-NOS3 Mouse Monoclonal Antibody
Antibodies
Western Blot: 2 ?g/ml; ELISA: 0.1 ?g/ml

Type: Primary
Antigen: NOS3 (nitric oxide synthase 3 (endothelial cell))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR