Anti-COL6 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26861-0.5EA 0 EUR
BIRBORB26861-0.5
Anti-COL6 Rabbit Polyclonal Antibody
Antibodies
DB: 1:5000-1:10000; ELISA: 1:4000-1:8000; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: COL6 (collagen, type VI)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR