Anti-COL1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB26866-0.5EA 0 EUR
BIRBORB26866-0.5
Anti-COL1 Rabbit Polyclonal Antibody
Antibodies
Immunofluorescence: 1:40; ELISA: 1:10-1:200

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR