Anti-Methylglyoxal-AGE Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB27265-30EA 0 EUR
BIRBORB27265-30
Anti-Methylglyoxal-AGE Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Methylglyoxal-AGE
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR