Anti-KDR Mouse Monoclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB27270-50EA 0 EUR
BIRBORB27270-50
Anti-KDR Mouse Monoclonal Antibody (Biotin)
Antibodies
ELISA: 1-15 ?g/ml; Western Blot 1-2 ?g/ml; FACS 2-5 ?g/ml

Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR