Anti-alpha Interleukin 1 Mouse Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27366-50EA 0 EUR
BIRBORB27366-50
Anti-alpha Interleukin 1 Mouse Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:1000; Immunochemistry: 1:400

Type: Primary
Antigen: alpha Interleukin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR