Anti-Cytokeratin, Hair Cortex Mouse monoclonal antibody

Supplier: Biorbyt
BIRBORB27367-0.1EA 0 EUR
BIRBORB27367-0.1
Anti-Cytokeratin, Hair Cortex Mouse monoclonal antibody
Antibodies
Western Blot: 1:500-1:1500

Type: Primary
Antigen: Cytokeratin, Hair Cortex
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR