Imprimer

Anti-MKI67 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB27421-5EA 0 EUR
BIRBORB27421-5
Anti-MKI67 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR