Anti-IL6 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27565-100EA 0 EUR
BIRBORB27565-100
Anti-IL6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL6 (interleukin 6 (interferon, beta 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR