Anti-FGF7 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27583-100EA 0 EUR
BIRBORB27583-100
Anti-FGF7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGF7 (fibroblast growth factor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR