Anti-EGF Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB27610-100EA 0 EUR
BIRBORB27610-100
Anti-EGF Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: EGF (epidermal growth factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR