Anti-PIK3R1 None Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27619-100EA 0 EUR
BIRBORB27619-100
Anti-PIK3R1 None Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PIK3R1 (phosphoinositide-3-kinase, regulatory subunit 1 (alpha))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR