Anti-CXCL13 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27639-100EA 0 EUR
BIRBORB27639-100
Anti-CXCL13 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CXCL13 (chemokine (C-X-C motif) ligand 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR