Anti-FABP2 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB27670-100EA 0 EUR
BIRBORB27670-100
Anti-FABP2 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: FABP2 (fatty acid binding protein 2, intestinal)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR