Anti-CEBPA Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB27675-100EA 0 EUR
BIRBORB27675-100
Anti-CEBPA Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: CEBPA (CCAAT/enhancer binding protein (C/EBP), alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR