Anti-IgM Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27702-0.25EA 0 EUR
BIRBORB27702-0.25 BIRBORB27702-0.5
Anti-IgM Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR