Anti-IgG Rabbit Antibody

Supplier: Biorbyt
BIRBORB27707-0.5EA 0 EUR
BIRBORB27707-0.5
Anti-IgG Rabbit Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR