Anti-Lipocalin 2 protein Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB27755-100EA 0 EUR
BIRBORB27755-100
Anti-Lipocalin 2 protein Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Lipocalin 2 protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR