Anti-ADIPOQ Rabbit Polyclonal Antibody
Supplier: Biorbyt
BIRBORB27760-100EA 0 EUR
BIRBORB27760-100
Anti-ADIPOQ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADIPOQ (adiponectin, C1Q and collagen domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human