Anti-Retinol binding protein 4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27776-100EA 0 EUR
BIRBORB27776-100
Anti-Retinol binding protein 4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Retinol binding protein 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR