Anti-FGF19 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB27780-100EA 0 EUR
BIRBORB27780-100
Anti-FGF19 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR