Anti-ADORA2A Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40781-100EA 0 EUR
BIRBORB40781-100
Anti-ADORA2A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-1:1000

Type: Primary
Antigen: ADORA2A (adenosine A2a receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR