Anti-FFAR4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40785-100EA 0 EUR
BIRBORB40785-100
Anti-FFAR4 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:500

Type: Primary
Antigen: FFAR4 (free fatty acid receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR