Imprimer

Anti-FLNB Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB40822-100EA 0 EUR
BIRBORB40822-100
Anti-FLNB Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: FLNB (filamin B, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR