Anti-APP Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40829-100EA 0 EUR
BIRBORB40829-100
Anti-APP Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:500-1:3000

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR