Imprimer

Anti-TTC1 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB40855-100EA 0 EUR
BIRBORB40855-100
Anti-TTC1 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000

Type: Primary
Antigen: TTC1 (tetratricopeptide repeat domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR