Anti-GABRB1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB40983-100EA 0 EUR
BIRBORB40983-100
Anti-GABRB1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: GABRB1 (gamma-aminobutyric acid (GABA) A receptor, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR