Imprimer

Anti-PAPD5 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41240-100EA 0 EUR
BIRBORB41240-100
Anti-PAPD5 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000

Type: Primary
Antigen: PAPD5 (PAP associated domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR