Anti-FAM50A Rabbit Polyclonal Antibody
Supplier: Biorbyt
BIRBORB41279-100EA 0 EUR
BIRBORB41279-100
Anti-FAM50A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000
Type: Primary
Antigen: FAM50A (family with sequence similarity 50, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human