Anti-FAM9B Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41281-100EA 0 EUR
BIRBORB41281-100
Anti-FAM9B Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: FAM9B (family with sequence similarity 9, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR