Anti-MRGPRX4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41423-100EA 0 EUR
BIRBORB41423-100
Anti-MRGPRX4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: MRGPRX4 (MAS-related GPR, member X4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR