Anti-I 309 Rabbit Polyclonal Antibody
Supplier: Biorbyt
BIRBORB41446-100EA 0 EUR
BIRBORB41446-100
Anti-I 309 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000
Type: Primary
Antigen: I 309
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human