Anti-CBFA2T2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41491-100EA 0 EUR
BIRBORB41491-100
Anti-CBFA2T2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: CBFA2T2 (core-binding factor, runt domain, alpha subunit 2; translocated to, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR