Imprimer

Anti-RAG2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41559-100EA 0 EUR
BIRBORB41559-100
Anti-RAG2 Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: RAG2 (recombination activating gene 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR