Imprimer

Anti-VAPA Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB41705-100EA 0 EUR
BIRBORB41705-100
Anti-VAPA Rabbit Polyclonal Antibody
Anticorps
Western Blot: 1:500-1:3000

Type: Primary
Antigen: VAPA (VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR