Anti-NDUFB5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41709-100EA 0 EUR
BIRBORB41709-100
Anti-NDUFB5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: NDUFB5 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR