Anti-DLST Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB41757-100EA 0 EUR
BIRBORB41757-100
Anti-DLST Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:3000

Type: Primary
Antigen: DLST (dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR