Anti-YWHAQ Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43278-100EA 0 EUR
BIRBORB43278-100
Anti-YWHAQ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: YWHAQ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR