Imprimer

Anti-IgG Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB43286-100EA 0 EUR
BIRBORB43286-100
Anti-IgG Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR