Anti-PROX1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43377-100EA 0 EUR
BIRBORB43377-100
Anti-PROX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PROX1 (prospero homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR