Anti-FBXL3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43401-100EA 0 EUR
BIRBORB43401-100
Anti-FBXL3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FBXL3 (F-box and leucine-rich repeat protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR