Anti-CDH1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43407-100EA 0 EUR
BIRBORB43407-100
Anti-CDH1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR