Anti-PECAM1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB43421-100EA 0 EUR
BIRBORB43421-100
Anti-PECAM1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR